Nuclear receptor ligand library plates, SCREEN-WELL®
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BML28020100 BML28020500
ENZOBML28020100EA 2220 EUR
ENZOBML28020100 ENZOBML28020500
Nuclear receptor ligand library plates, SCREEN-WELL®
Microplaten HTS-microplaten
The SCREEN-WELL® Nuclear Receptor Ligand Library contains 74 compounds with defined, putative and potential activity at nuclear receptors.
  • Stable for one year from the date of receipt when stored at –80 °C
  • Shipped on dry ice
  • Receptor agonists and antagonists are included
The library consists of a chemically diverse group of compounds which are supplied dissolved in DMSO at 10 mM (1 mM for 5 compounds) and aliquoted into deep-wel plates. The library is an ideal tool for chemical genomics, receptor de-orphaning and other routine pharmacologicalMore Product Information
Order Now
Specificaties
Environmentally Preferable