L(+)-Arginine
Leverancier: Serva
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
H-L-Arg-OH, (S)-2-Amino-5-guanidinopentanoic acid, Arginine, (S)-(+)-Arginine, L-Arginine, H-Arg-OH
13909.02
13909.03
SERA13909.02EA
29
EUR
SERA13909.02
SERA13909.03
L(+)-Arginine
L(+)-Arginine
Formule:
C₆H₁₄N₄O₂ Kookpunt: 368 °C (1013 hPa) Smeltpunt: 238 °C Dichtheid: 1,3 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00002635 CAS nummer: 74-79-3 EINECS: 200-811-1 Merck Index: 13,00785 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...