VWR®, Yeast nitrogen base (YNB)
Leverancier: VWR ChemicalsTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
Yeast Nitrogen Base (YNB) is a base medium for preparation of minimal and synthetic defined yeast media.
- Animal origin
- Suitable for studying strains of yeast requiring certain vitamins
Bestelinformatie: Base materials are available with or without ammonium sulfate for the classification of yeast.