L-Arginine hydrochloride ≥99% (by titrimetric analysis), Millipore®
Leverancier: Merck Millipore Calbiochem
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: H-L-Arg-OH.HCl, (S)-2-Amino-5-guanidinopentanoic acid hydrochloride, (S)-(+)-Arginine HCl, H-Arg-OH·HCl, L-Arginine, hydrochloride, (S)-(+)-Arginine hydrochloride, H-Arg-OH.HCl, L-Arg, L(+)-Arginine monohydrochloride, L(+)-Arginine hydrochloride
181003-1 181003-50
181003-1EA 609 EUR
181003-1 181003-50
L-Arginine hydrochloride ≥99% (by titrimetric analysis), Millipore®
L-Arginine hydrochloride
Formule: C₆H₁₅ClN₄O₂
MW: 210.66 g/mol
Smeltpunt: 218
Dichtheid: 1.42 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064550
CAS nummer: 1119-34-2
EINECS: 214-275-1
Merck Index: 13,00785