L(+)-Cysteine for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
M-Clarity™ quality level = MQ200
8.16014.0025 8.16014.0100
8.16014.0025EA 59.2 EUR
8.16014.0025 8.16014.0100
L(+)-Cysteine for synthesis, Sigma-Aldrich®
(R)-(+)-Cysteine
Formule: HSCH₂CH(NH₂)CO₂H
Kookpunt: 254 °C (1013 hPa)
Smeltpunt: 115 °C
Dichtheid: 1.523 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064306
CAS nummer: 52-90-4
EINECS: 200-158-2
Merck Index: 13,02810
Order Now
Specification Test Results
Solubility 160 g/l (20 °C)
Melting Point 220 - 228 °C
Molar Mass 121.16 g/mol
Bulk Density 300 kg/m3
pH 4.5 - 5.5 (100 g/l, H2O, 20 °C)
Ignition Point 420 °C