L(+)-Aspartic acid, Millipore®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: (S)-(+)-Aminosuccinic acid (S)-Aminobutanedioic acid (S)-Aminosuccinic acid H-Asp-OH L-Asparaginic acid L(+)-Aspartic acid, (S)-(+)-Aspartic acid, L(+)-2-Amino-1,4-butanedioic acid, (S)-(+)-2-Amino-1,4-butanedioic acid, L-Aspartic acid, H-Asp-OH, L-α-Aminosuccinic acid, L-αlpha-Aminosuccinic acid, (S)-2-Aminosuccinic acid, Asp, D
MQ100
1.00126.1000 1.00126.9010
1.00126.1000EA 334 EUR
1.00126.1000 1.00126.9010
L(+)-Aspartic acid, Millipore®
L(+)-Aspartic acid
Soluble in water and mineral acids; insoluble in organic solvents.
Formule: C₄H₇NO₄
MW: 133.1 g/mol
Kookpunt: 311
Smeltpunt: 251
Dichtheid: 1.6613 g/cm³ (13 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002616
CAS nummer: 56-84-8
EINECS: 200-291-6
Merck Index: 13,00845