L(+)-Aspartic acid

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: (S)-(+)-Aminosuccinic acid (S)-Aminobutanedioic acid (S)-Aminosuccinic acid H-Asp-OH L-Asparaginic acid L(+)-Aspartic acid, (S)-(+)-Aspartic acid, L(+)-2-Amino-1,4-butanedioic acid, (S)-(+)-2-Amino-1,4-butanedioic acid, L-Aspartic acid, H-Asp-OH, L-α-Aminosuccinic acid, L-αlpha-Aminosuccinic acid, (S)-2-Aminosuccinic acid, Asp, D

T202.1 T202.2 T202.3
ROTHT202.1EA 28.5 EUR
ROTHT202.1 ROTHT202.2 ROTHT202.3
L(+)-Aspartic acid
L(+)-Aspartic acid
Formule: C₄H₇NO₄
MW: 133.1 g/mol
Kookpunt: 311
Smeltpunt: 251
Dichtheid: 1.6613 g/cm³ (13 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002616
CAS nummer: 56-84-8
EINECS: 200-291-6
Merck Index: 13,00845

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR