L(+)-Aspartic acid
Leverancier: Serva
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
(S)-(+)-Aminosuccinic acid (S)-Aminobutanedioic acid (S)-Aminosuccinic acid H-Asp-OH L-Asparaginic acid L(+)-Aspartic acid, (S)-(+)-Aspartic acid, L(+)-2-Amino-1,4-butanedioic acid, (S)-(+)-2-Amino-1,4-butanedioic acid, L-Aspartic acid, H-Asp-OH, L-α-Aminosuccinic acid, L-αlpha-Aminosuccinic acid, (S)-2-Aminosuccinic acid, Asp, D
14180.02
14180.03
SERA14180.02EA
53
EUR
SERA14180.02
SERA14180.03
L(+)-Aspartic acid
L(+)-Aspartic acid
Formule:
C₄H₇NO₄ MW: 133.1 g/mol Kookpunt: 311 Smeltpunt: 251 Dichtheid: 1.6613 g/cm³ (13 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00002616 CAS nummer: 56-84-8 EINECS: 200-291-6 Merck Index: 13,00845 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...