D-(+)-Malic acid 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: (R)-(+)-Hydroxysuccinic acid, D(+)-Hydroxy-1,4-butanedioic acid, (R)-(+)-Hydroxy-1,4-butanedioic acid, D(+)-Malic acid, D(+)-Hydroxysuccinic acid, (R)-2-Hydroxysuccinic acid
153500050 153500010 153500250
ACRO153500050EA 74.7 EUR
ACRO153500050 ACRO153500010 ACRO153500250
D-(+)-Malic acid 98%
D-(+)-Malic acid
Formule: C₄H₆O₅
MW: 134.09 g/mol
Kookpunt: 322
Smeltpunt: 99…104 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004245
CAS nummer: 636-61-3
EINECS: 211-262-2
Merck Index: 13,05307
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 99 to 104 °C
Water ≤1 % (K.F.)
Sulfated ash ≤0.1 %
HPLC ≤1 % Fumaric acid
Specific optical rotation +1.8° to +2.6° (20 °C, 589 nm) (c=3, Water)
Titration with NaOH ≥97.5 %