ESD occupational shoes, clogs, C-FIT, Michel
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
972430/41 972430/42 972430/40 972430/43 972430/44 972430/45 972430/46 972430/47 972430/48 972440/40 972440/41 972440/42 972440/43 972440/44 972440/45 972440/46 972440/47 972440/48
ELTN972430/41EA 54.9 EUR
ELTN972430/41 ELTN972430/42 ELTN972430/40 ELTN972430/43 ELTN972430/44 ELTN972430/45 ELTN972430/46 ELTN972430/47 ELTN972430/48 ELTN972440/40 ELTN972440/41 ELTN972440/42 ELTN972440/43 ELTN972440/44 ELTN972440/45 ELTN972440/46 ELTN972440/47 ELTN972440/48
ESD occupational shoes, clogs, C-FIT, Michel
Schoenen en laarzen
Metal-free clogs made from smooth, tear resistant microfibre upper, an adjustable Velcro® heel strap, and a mono-density PU sole (2,5 mm, oil, fuel and abrasion resistant, heat resistant up to approx. 130 °C).
  • Climate regulating, breathable textile lining
  • Full length inlay sole (C-Fit ESD)
  • ESD soft fleece insole
  • Anatomically-shaped insole and ankle support for improved stability
  • Shock-absorbent heel
Colour: Black or white
Certificaten: EN 61340-5-1 (ESD); EN ISO 20347; OB, SRC, A, E, FO.