ESD occupational shoes, clogs, C-FIT, Mia
Leverancier: ELTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
974430/35 974430/36 974430/37 974430/38 974430/39 974430/40 974430/41 974430/42 974440/35 974440/36 974440/37 974440/38 974440/39 974440/40 974440/41 974440/42
ELTN974430/35EA 54.9 EUR
ELTN974430/35 ELTN974430/36 ELTN974430/37 ELTN974430/38 ELTN974430/39 ELTN974430/40 ELTN974430/41 ELTN974430/42 ELTN974440/35 ELTN974440/36 ELTN974440/37 ELTN974440/38 ELTN974440/39 ELTN974440/40 ELTN974440/41 ELTN974440/42
ESD occupational shoes, clogs, C-FIT, Mia
Schoenen en laarzen
Metal-free clogs made from smooth, tear resistant microfibre upper, an adjustable Velcro® heel strap, and a mono-density PU sole (2,5 mm, oil, fuel and abrasion resistant, heat resistant up to approx. 130 °C).
  • Climate regulating, breathable textile lining
  • Full-length inlay sole C-FIT ESD
  • ESD soft fleece insole
  • Anatomically shaped insole and ankle support for improved stability
  • Shock absorber heel
The shoe is tailored to the ergonomics of female feet.
Colour: Black or white
Certificaten: EN 61340-5-1 (ESD); EN ISO 20347; OB, SRC, A, E, FO.