D-(-)-Valine

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: H-D-Val-OH, (R)-(-)-Valine

4031330.0025 4031330.0100
BACE4031330.0025EA 0 EUR
BACE4031330.0025 BACE4031330.0100
D-(-)-Valine
D-(-)-Valine
Formule: C₅H₁₁NO₂
Kookpunt: 217 °C (1013 hPa)
Smeltpunt: 86 °C
MDL Nummer: MFCD00064219
CAS nummer: 640-68-6
EINECS: 211-368-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR