D-(-)-Histidine

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: (R)-(-)-Histidine, D-3-Imidazol-4-ylalanine, (2R)-(-)-Histidine, H-D-His-OH

4030626.0005 4030626.0025 4030626.0100
BACE4030626.0005EA 0 EUR
BACE4030626.0005 BACE4030626.0025 BACE4030626.0100
D-(-)-Histidine
D-(-)-Histidine
Formule: C₆H₉N₃O₂
MW: 155.16 g/mol
Smeltpunt: 287
MDL Nummer: MFCD00065963
CAS nummer: 351-50-8
EINECS: 206-513-8
Merck Index: 12,04758

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR