D-(+)-Tyrosine

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

4026808.0005 4026808.0025 4026808.0100
BACE4026808.0005EA 0 EUR
BACE4026808.0005 BACE4026808.0025 BACE4026808.0100
D-(+)-Tyrosine
D-(+)-Tyrosine
Formule: C₉H₁₁NO₃
Kookpunt: 351 °C (1013 hPa)
Smeltpunt: 285 °C
MDL Nummer: MFCD00063073
CAS nummer: 556-02-5
EINECS: 209-112-6
Merck Index: 12,09970

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR