L(-)-Tyrosine

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: L-Tyrosine, (S)-(-)-Tyrosine, Tyrosine, H-Tyr-OH

4031323.0250 4031323.1000
BACE4031323.0250EA 0 EUR
BACE4031323.0250 BACE4031323.1000
L(-)-Tyrosine
L(-)-Tyrosine
Formule: C₉H₁₁NO₃
MW: 181.19 g/mol
Kookpunt: 351
Smeltpunt: 285
Dichtheid: 1.41 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002606
CAS nummer: 60-18-4
EINECS: 200-460-4
Merck Index: 13,09907

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR