(R)-(+)-α-Methylbenzylamine for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: (R)-(+)-α-Phenylethylamine, (R)-(+)-1-Phenylethylamine, D(+)-α-Methylbenzylamine
M-Clarity™ quality level = MQ200
8.07031.0010 8.07031.0100
8.07031.0010EA 29.6 EUR
8.07031.0010 8.07031.0100
(R)-(+)-α-Methylbenzylamine for synthesis, Sigma-Aldrich®
(R)-(+)-α-Methylbenzylamine
Formule: C₈H₁₁N
MW: 121.18 g/mol
Kookpunt: 180
Smeltpunt: 32.5
Dichtheid: 0.95 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064405
CAS nummer: 3886-69-9
EINECS: 223-423-4
UN: 2922
ADR: 8 (6.1) I
Merck Index: 12,06106
Order Now
Specification Test Results
Solubility 40 g/l (20 °C)
Melting Point -10 °C
Molar Mass 121.18 g/mol
Boiling Point 187 °C (1013 hPa)
Vapor Pressure 0.4 hPa (20 °C)
Flash Point 71 °C DIN 51758
Refractive Index 1.5265 (24 °C, 589 nm)
Density 0.95 g/cm3 (20 °C)
pH 7 (40 g/l, H2O, 20 °C)
Ignition Point 355 °C DIN 51794