1,10-Phenanthroline monohydrate

Leverancier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

9113.1 9113.2
ROTH9113.1EA 0 EUR
ROTH9113.1 ROTH9113.2
1,10-Phenanthroline monohydrate
1,10-Phenanthroline monohydrate
Formule: C₁₂H₈N₂·1H₂O
Smeltpunt: 93…94 °C
Dichtheid: 1,259 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149973
CAS nummer: 5144-89-8
EINECS: 200-629-2
UN: 2811
ADR: 6.1,III
Merck Index: 13,07291
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aba346b9f67fdda9afd4d48b893e6318564d604c&vid=2d08e04ee9525293a747b55cdcff8f66a6c4715a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR