Sorbic acid, EMPROVE® ESSENTIAL FCC, BP, Ph. Eur., E200, NF suitable for use as excipient, SAFC®
Leverancier: MERCK PRODUCTION CHEMICALS
Ratings:
Avg. Ratings: 5.0 (1 Review)
Total Ratings: 1
Avg. Ratings: 5.0 out of 5
Was this review helpful? Yes No
Yes: 0 No: 0 
food pastry industry / produit / constantine algerie
Yes, I recommend
Acide sorbique, EMPROVE® ESSENTIAL Ph. Eur., BP, NF, FCC, E200
Waarschuwing
Synoniemen: (E,E)-2,4-Hexadienoic acid, (2E,4E)-2,4-Hexadienoic acid, trans,trans-2,4-Hexadienoic acid, 2,4-Hexadienoic acid, (2E,4E)-Hexa-2,4-dienoic acid
1.00662.5000 1.00662.9025
1.00662.5000EA 595 EUR
1.00662.5000 1.00662.9025
Sorbic acid, EMPROVE® ESSENTIAL FCC, BP, Ph. Eur., E200, NF suitable for use as excipient, SAFC®
Sorbic acid
Formule: CH₃CH=CHCH=CHCOOH
Kookpunt: 228 °C (1013 hPa)
Smeltpunt: 132…135 °C
Dichtheid: 1,204 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002703
CAS nummer: 110-44-1
EINECS: 203-768-7
Merck Index: 13,08793
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=440c50b4fc52d9ff0b52afa7ba596c860f95e0e7&vid=f9b0ff409010208781696e9ca3314c02aaf1af36&prid=web&clid=SAPDAM