4-Bromobutyric acid 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
133470050 133470250 133471000
ACRO133470050EA 42.9 EUR
ACRO133470050 ACRO133470250 ACRO133471000
4-Bromobutyric acid 98%
4-Bromobutyric acid
Formule: BrH₂C(CH₂)₂COOH
Kookpunt: 249 °C (1013 hPa)
Smeltpunt: 28…32 °C
Vlampunt: 124 °C
MDL Nummer: MFCD00002817
CAS nummer: 2623-87-2
EINECS: 220-083-9
UN: 3261
ADR: 8,III
Order Now
Specification Test Results
Appearance Light brown Low melting crystals
Infrared spectrum Conforms
Melting point 28 to 32 °C
Titration with NaOH 97.5 to 102.5 %