5-Bromovaleric acid for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 5-Bromopentanoic acid
M-Clarity™ quality level = MQ200
8.18329.0050 8.18329.0250
8.18329.0050EA 92.8 EUR
8.18329.0050 8.18329.0250
5-Bromovaleric acid for synthesis, Sigma-Aldrich®
5-Bromovaleric acid
Formule: BrH₂C(CH₂)₃COOH
MW: 181.03 g/mol
Kookpunt: 266
Smeltpunt: 38…40 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004414
CAS nummer: 2067-33-6
EINECS: 218-185-3
UN: 3261
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 38 - 41 °C
Molar Mass 181.02 g/mol
Boiling Point 142 - 145 °C (17 hPa)
Flash Point 112 °C