Trichloroacetamide 98+%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
202920250
202921000
202925000
ACRO202920250EA
0
EUR
ACRO202920250
ACRO202921000
ACRO202925000
Trichloroacetamide 98+%
Trichloroacetamide
Formule:
C₂H₂Cl₃NO Kookpunt: 238…240 °C (760 mmHg) Smeltpunt: 139…143 °C Dichtheid: 1,82 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00008009 CAS nummer: 594-65-0 EINECS: 209-849-3 |
Specification Test Results
Appearance | White to off-white Powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 139 to 143 °C |
HPLC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...