Sebaconitrile 95%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
1,8-Dicyanooctane, Sebacic acid dinitrile
B23010.06
B23010.14
B23010.06EA
23.1
EUR
B23010.06
B23010.14
Sebaconitrile 95%
Sebaconitrile
Purity: 95%
CAS No.: 1871-96-1
Molecular Formula: NC(CH2)8CN
Formula Weight: 164.25
Flash Point:
Formule:
C₁₀H₁₆N₂ Kookpunt: 199…200 °C Smeltpunt: 4…8 °C Dichtheid: 0,910 Vlampunt: >110 °C (230 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00019900 CAS nummer: 1871-96-1 EINECS: 217-494-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...