Di-tert-butyl pyrocarbonate 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Boc Anhydride, Di-tert-butyl dicarbonate, DIBOC
194670100 194670250 194670500 194671000 194675000
ACRO194670100EA 45.2 EUR
ACRO194670100 ACRO194670250 ACRO194670500 ACRO194671000 ACRO194675000
Di-tert-butyl pyrocarbonate 99%
Di-tert-butyl pyrocarbonate
Formule: C₁₀H₁₈O₅
Kookpunt: 56…57 °C (,5 mmHg)
Smeltpunt: 22…24 °C
Dichtheid: 1,02 g/cm³
Vlampunt: 37 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00008805
CAS nummer: 24424-99-5
EINECS: 246-240-1
UN: 2930
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae68e9d9dc1f713c2e5ac96bd05040cec5c7bcdd&vid=719e05eb5ef4a6a4176af5e827ff4390d03d64ab&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White Low melting crystalline solid
Infrared spectrum Conforms
Titration by coupling 98.5 to 101.5 %