Di-tert-butyl pyrocarbonate ≥97%
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Boc Anhydride, Di-tert-butyl dicarbonate, DIBOC
Purity: 97+%
CAS No.: 24424-99-5
Molecular Formula: C10H18O5
Formula Weight: 218.25
Storage Temperature: 2°C to 8°C
Flash Point: 37°(99°F)
Physical Form: Low-melting solid
Density: 0.950
Refractive Index: 1.4090
Sensitivity: Moisture Sensitive
Appearance: White to pale yellow
Odour: Mild
Melting Point: 21-24°
Boiling Point: 56-57°/0.5mm
Beilstein Reference: 1911173
UN No.: 2930
MDL No.: MFCD00008805
Reagent for the introduction of the Boc protecting group
Formule:
((H₃C)₃COC(O))₂O Kookpunt: 56…57 °C (0,5 mmHg) Smeltpunt: 21…24 °C Dichtheid: 0,950 Vlampunt: 37 °C (99 °F) Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00008805 CAS nummer: 24424-99-5 EINECS: 246-240-1 UN: 2930 ADR: 6.1,II |