Diethyl pyrocarbonate 97,5, high_purity
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: DEPC (Diethylpyrocarbonate), Diethyl dicarbonate, DEPC
E174-25GEA 98.8 EUR
E174-25G E174-100G
Diethyl pyrocarbonate 97,5, high_purity
Diethyl pyrocarbonate
DEPC is used to inactivate RNase enzymes from water and other laboratory utensils. DEPC covalently modifies histidine residues in RNase enzymes. DEPC can be used with PBS or MOPS buffer, but is incompatible with Tris and HEPES.
Formule: O(COOC₂H₅)₂
MW: 162.14 g/mol
Kookpunt: 93 – 94 °C
Dichtheid: 1.123 g/cm³ (20 °C)
Vlampunt: 69
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00009106
CAS nummer: 1609-47-8
EINECS: 216-542-8
Merck Index: 13,08088
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9702c5e35cd698e404dd5a1f3fdc66f22333ae6a&vid=2b31173ec3b52910da8dcf5c40d7e8be6f3817df&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity (acid assay) >97.5%