Diethyl pyrocarbonate

Leverancier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: DEPC (Diethylpyrocarbonate), Diethyl dicarbonate, DEPC

18835.01 18835.02
SERA18835.01EA 62 EUR
SERA18835.01 SERA18835.02
Diethyl pyrocarbonate
Diethyl pyrocarbonate
Formule: O(COOC₂H₅)₂
MW: 162.14 g/mol
Kookpunt: 93
Dichtheid: 1.123 g/cm³ (20 °C)
Vlampunt: 69
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00009106
CAS nummer: 1609-47-8
EINECS: 216-542-8
Merck Index: 13,08088

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR