4-Oxopimelic acid 98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
122880050
122880250
122881000
ACRO122880050EA
64.3
EUR
ACRO122880050
ACRO122880250
ACRO122881000
4-Oxopimelic acid 98%
4-Oxopimelic acid
Formule:
C₇H₁₀O₅ Smeltpunt: 142…144 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00002798 CAS nummer: 502-50-1 EINECS: 207-941-8 |
Specification Test Results
Appearance | White to yellow Powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 141 to 145 °C |
Titration with NaOH | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...