N,N-Dimethylformamide, anhydrous 99,8
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
Argon_Chemseal
43997.K2
43997.M1
43997.K2EA
243
EUR
43997.K2
43997.M1
N,N-Dimethylformamide, anhydrous 99,8
N,N-Dimethylformamide
Purity: 99.8%
CAS No.: 68-12-2
Molecular Formula: HCON(CH3)2
Formula Weight: 73.09
Flash Point: 57°(135°F)
Physical Form: Liquid
Density: 0.944
Refractive Index: 1.4310
Sensitivity: Hygroscopic
Odour: Amine-like
pH: 7
Melting Point: -61°
Boiling Point: 153°
Beilstein Reference: 605365
Merck Reference: 14,3243
UN No.: 2265
MDL No.: MFCD00003284
Formule:
HCON(CH₃)₂ MW: 73.09 g/mol Kookpunt: 153 °C Smeltpunt: –61 °C Dichtheid: 0.944 Vlampunt: 57 °C (135 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00003284 CAS nummer: 68-12-2 EINECS: 200-679-5 UN: 2265 ADR: 3 III Bereik: 01-2119475605-32 Merck Index: 14,3243 |