Electroporation system, high throughput ECM™ 830
Leverancier: BTX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
45-0665 45-0666
BTXI45-0665EA 21630 EUR
BTXI45-0665 BTXI45-0666
Electroporation system, high throughput ECM™ 830
Elektroporatiesystemen Elektroporatoren
The High Throughput ECM™ 830 system offers a multi-well electroporation technology for processing multiple samples in seconds. Using the HT multi-well plates instead of traditional cuvettes, the researcher can transition from a single cuvette to either a 25-well or 96-well electroporation plate by using one simple plate handler. This increases yields and the number of experiments run in a single day. Experiments take seconds to run, allowing for quick and efficient optimisation of the electrical and biological parameters. Once optimised, samples are rapidly processed increasing yields and saving valuable time and money.
  • Can be used in mammalian cell transfection, including primary and stem cells
  • Screening of cDNA and siRNA library
  • High throughput 25 to 96-well
The key to the high throughput system is the addition of a plate handler and multi-well electroporation plates. The HT-100 and HT-200 Plate Handlers have pulse switching technology integrated into the package and gold-plated contacts to mate with a 25- or 96-well electroporation plate. The plate handler delivers a single pulse to the wells of the plate, column by column, using parameters set in the ECM™ 830 generator. The HT-100 uses manual track switching, while the HT-200 employs auto-sense track switching. Both plate handlers work with 96-well plates and come with an adapter to accommodate 25-well plates with our ECM™ generator.
* Includes ECM™ 830 generator, 25-well plates (2 mm gap, 6X), plate seals, HT-100/HT-200 plate handler and a plate adaptor.
** Includes ECM™ 830 generator, 95-well plates (2 mm gap, 6X), plate seals, HT-100/HT-200 plate handler and a plate adaptor.
*** Includes ECM™ 830 generator, 630B safety stand, cuvettes 1 mm, 2 mm, 4 mm package of 30 (10 each), cuvette rack 660, enhancer 3000 probe, enhancer interface box, oscilloscope, cables, 25-well plates (2 mm gap, 6X), HT-200 plate handler and a plate adaptor.