N,N-Dimethylformamide
Leverancier: Roth Carl
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
T921.1
T921.2
ROTHT921.1EA
68.5
EUR
ROTHT921.1
ROTHT921.2
N,N-Dimethylformamide
N,N-Dimethylformamide
Formule:
HCON(CH₃)₂ Kookpunt: 153 °C (1013 hPa) Smeltpunt: –61 °C Dichtheid: 0,949 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00003284 CAS nummer: 68-12-2 EINECS: 200-679-5 UN: 2265 ADR: 3,III Bereik: 01-2119475605-32 Merck Index: 13,03269 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...