Formamide ≥99.5% ACS
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Carbamaldehyde
14835.30
14835.A1
14835.D6
14835.30EA
76.3
EUR
14835.30
14835.A1
14835.D6
Formamide ≥99.5% ACS
Formamide
Purity: 99.5+%
CAS No.: 75-12-7
Molecular Formula: CH3NO
Formula Weight: 45.04
Flash Point: 154°(310°F)
Physical Form: Liquid
Density: 1.134
Refractive Index: 1.4475
Sensitivity: Hygroscopic
Appearance: Colourless
Odour: Ammonia-like
Melting Point: 2°
Boiling Point: 210°
Beilstein Reference: 505995
Merck Reference: 14,4237
MDL No.: MFCD00007941
Formule:
HCONH₂ Kookpunt: 210 °C Smeltpunt: 2 °C Dichtheid: 1,134 Vlampunt: 154 °C (310 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00007941 CAS nummer: 75-12-7 EINECS: 200-842-0 Merck Index: 14,4237 |