Ureum ≥98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
A12360.0C A12360.0E A12360.36
A12360.0CEA 118 EUR
A12360.0C A12360.0E A12360.36
Ureum ≥98%
Ureum
Purity: 98+% CAS No.: 57-13-6 Molecular Formula: NH2CONH2 Formula Weight: 60.06 Odour: Ammonia-like pH: 9,2-9,5 Melting Point: 130-135° Beilstein Reference: 635724 Merck Reference: 14,9867 MDL No.: MFCD00008022
Formule: NH₂CONH₂
Kookpunt: 197 °C (1013 hPa)
Smeltpunt: 130…135 °C
Dichtheid: 1,335
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008022
CAS nummer: 57-13-6
EINECS: 200-315-5
Bereik: 01-2119463277-33
Merck Index: 14,9867
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM