Methyl butyrate, Supelco®
Leverancier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Butyric acid methyl ester, Butanoic acid methyl ester
M-Clarity™ quality level = MQ100
19358-1ML
19358-5ML
19358-1MLEA
54.3
EUR
19358-1ML
19358-5ML
Methyl butyrate, Supelco®
Methyl butyrate
Formule:
C₅H₁₀O₂ MW: 102.13 g/mol Kookpunt: 102 Smeltpunt: –84 °C Dichtheid: 0.8984 g/cm³ (20 °C) Vlampunt: 14 |
MDL Nummer:
MFCD00009391 CAS nummer: 623-42-7 EINECS: 210-792-1 UN: 1237 ADR: 3 II Merck Index: 13,06062 |