Chemiluminescent acridinium esters
Leverancier: Enzo Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ENZOADI906001EA 201.3 EUR
ENZOADI906001 ENZOADI9060001 ALEXADI-906-001 ALEXADI-906-000-1
Chemiluminescent acridinium esters
Reagentia voor elektroforese Western Blotting Reagents
Chemiluminescent acridinium esters are commonly used to label antibodies, proteins, nucleic acids, and some peptides, and are frequently used as detection reagents in immunoassays.
  • Species independent