Supelco® Organic Reference Standard, Methyl Myristate
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
70129-1ML 70129-5ML
70129-1MLEA 50 EUR
70129-1ML 70129-5ML
Supelco® Organic Reference Standard, Methyl Myristate
Standaarden Organische standaarden
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3816abc8582d4b1aabde511da753ba408c55f921&vid=3816abc8582d4b1aabde511da753ba408c55f921&prid=web&clid=SAPDAM
Order Now
Specificaties
Synonym(s) Methyl tetradecanoate; Myristic acid methyl ester
M-Clarity quality level MQ100
Environmentally Preferable
Dichtheid 0,855 g/ml at 25 °C (lit.)
CAS No. 124-10-7
MDL No. MFCD00008983
Formule CH₃(CH₂)₁₂COOCH₃
Moleculair gewicht 242,40
Smeltpuntapparaat 18 °C (lit.)
Kookpunt 323 °C (lit.)
Vlampunt 112,0 °C - closed cup
Shipping conditions STD
Storage conditions RT