Diethylcarbonaat 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
114220010 114220025 114225000 114220100 114221000
ACRO114220010EA 69.6 EUR
ACRO114220010 ACRO114220025 ACRO114225000 ACRO114220100 ACRO114221000
Diethylcarbonaat 99%
Diethylcarbonaat
Formule: C₅H₁₀O₃
MW: 118.13 g/mol
Kookpunt: 126…128 °C (760 mmHg)
Smeltpunt: –43 °C
Dichtheid: 0.975 g/cm³
Vlampunt: 33 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009107
CAS nummer: 105-58-8
EINECS: 203-311-1
UN: 2366
ADR: 3 III
Merck Index: 13,03815
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3830 to 1.3860 (20°C, 589 nm)
GC ≥98.5 %
Stabilizer Approx. 0.01 % Citric acid
Relative density (20/4°C) 1.026 to 1.036
Acid value ≤5 mg KOH/g