Diethylcarbonaat 99%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
A12477.0D
A12477.0F
A12477.AP
AB115877.0500
A12477.0DEA
323
EUR
A12477.0D
A12477.0F
A12477.AP
ABCRAB115877.0500
Diethylcarbonaat 99%
Diethylcarbonaat
Purity: 99+%
CAS No.: 105-58-8
Molecular Formula: (CH3CH2O)2C=O
Formula Weight: 118.13
Flash Point: 31°(87°F)
Physical Form: Clear liquid
Density: 0.975
Refractive Index: 1.3845
Sensitivity: Moisture Sensitive
Appearance: Colourless
Odour: Pleasant
Melting Point: -43°
Boiling Point: 126-128°
Beilstein Reference: 956591
Merck Reference: 14,3780
UN No.: 2366
MDL No.: MFCD00009107
Formule:
C₅H₁₀O₃ MW: 118.13 g/mol Kookpunt: 126…128 °C Smeltpunt: –43 °C Dichtheid: 0.975 Vlampunt: 31 °C (87 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00009107 CAS nummer: 105-58-8 EINECS: 203-311-1 UN: 2366 ADR: 3 III Merck Index: 13,03815 |