Diethyl succinate ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
|
Synoniemen:
Succinic acid diethyl ester, 1,4-Butanedioic acid diethyl ester
A15777.0C
A15777.22
A15777.36
A15777.0CEA
393
EUR
A15777.0C
A15777.22
A15777.36
Diethyl succinate ≥98%
Diethyl succinate
Purity: 98%
CAS No.: 123-25-1
Molecular Formula: C8H14O4
Formula Weight: 174.20
Flash Point: 90°(194°F)
Refractive Index: 1.4200
Melting Point: -21°
Boiling Point: 216-218°
Beilstein Reference: 907645
Merck Reference: 14,8869
MDL No.: MFCD00009208
Formule:
C₈H₁₄O₄ MW: 174.2 g/mol Kookpunt: 216…218 °C Smeltpunt: –21 °C Dichtheid: 1.043 Vlampunt: 90 °C (194 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00009208 CAS nummer: 123-25-1 EINECS: 204-612-0 Merck Index: 12,09038 |