Dimethyloxalaat ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

A15359.0E A15359.22 A15359.36
A15359.0EEA 304 EUR
A15359.0E A15359.22 A15359.36
Dimethyloxalaat ≥99%
Dimethyloxalaat

Purity: 99% CAS No.: 553-90-2 Molecular Formula: C4H6O4 Formula Weight: 118.09 Flash Point: 75°(167°F) Melting Point: 50-54° Boiling Point: 162-163° Beilstein Reference: 1071744 Merck Reference: 14,6106 MDL No.: MFCD00008442

Formule: C₄H₆O₄
Kookpunt: 162…163 °C
Smeltpunt: 50…54 °C
Dichtheid: 1,148
Vlampunt: 75 °C (167 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008442
CAS nummer: 553-90-2
EINECS: 209-053-6
UN: 3261
ADR: 8,III
Merck Index: 14,6106

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR