Octanoyl chloride 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
129431000 129435000 129430025
ACRO129431000EA 37.1 EUR
ACRO129431000 ACRO129435000 ACRO129430025
Octanoyl chloride 99%
Octanoyl chloride
Formule: C₈H₁₅ClO
MW: 162.66 g/mol
Kookpunt: 195 °C (760 mmHg)
Smeltpunt: –63 °C
Dichtheid: 0.95 g/cm³
Vlampunt: 75 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000766
CAS nummer: 111-64-8
EINECS: 203-891-6
UN: 2927
ADR: 6.1 II
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4340 to 1.4360 (20°C, 589 nm)
Titration Argentometric ≥98.5 %
GC ≥98.5 %
Color scale ≤80 APHA
Acidity ≤1 %
Phosphorus (P) ≤150 ppm