Lauric acid 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Dodecanoic acid, Dodecylic acid
167281000 167285000 167280050 167280010 167280051
ACRO167281000EA 35.5 EUR
ACRO167281000 ACRO167285000 ACRO167280050 ACRO167280010 ACRO167280051
Lauric acid 99%
Lauric acid
Formule: C₁₂H₂₄O₂
MW: 200.32 g/mol
Kookpunt: 225 °C (100 mmHg)
Smeltpunt: 44…46 °C
Vlampunt: 156 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002736
CAS nummer: 143-07-7
EINECS: 205-582-1
Merck Index: 13,05400
Order Now
Specification Test Results
Appearance White Crystalline powder or flakes
Infrared spectrum Conforms
GC ≥98.5 %