Natriumformiaat 99,0 ACS
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Formic acid sodium salt
36424.36
36424.A3
36424.36EA
66.7
EUR
36424.36
36424.A3
Natriumformiaat 99,0 ACS
Natriumformiaat
Purity: 99.0% min
CAS No.: 141-53-7
Molecular Formula: HCO2Na
Formula Weight: 68.01
Physical Form: Crystalline
Density: 1.92
Melting Point: 259-262°
Solubility: Soluble in water, glycerol. Slightly soluble in alcohol
Beilstein Reference: 3595134
Merck Reference: 14,8621
MDL No.: MFCD00013101
Reducing agent, buffering agent, complexing agent, Noble metal precipitant
Formule:
HCO₂Na MW: 68.01 g/mol Smeltpunt: 259…262 °C Dichtheid: 1.92 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00013101 CAS nummer: 141-53-7 EINECS: 205-488-0 Bereik: 01-2119486468-21 Merck Index: 14,8621 |