Sodium octanoate 96%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Octanoic acid sodium acid, Sodium caprylate, Caprylic acid sodium acid, Sodium n-Octanoate
A13451.22 A13451.36
A13451.22EA 52.7 EUR
A13451.22 A13451.36
Sodium octanoate 96%
Sodium octanoate
Purity: 96% CAS No.: 1984-06-1 Molecular Formula: CH3(CH2)6CO2Na Formula Weight: 166.20 Melting Point: 300° Beilstein Reference: 3597723
Formule: C₈H₁₅NaO₂
MW: 166.2 g/mol
Smeltpunt: >300 °C
Dichtheid: 0.9949 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00058511
CAS nummer: 1984-06-1
EINECS: 217-850-5