Zinc acetate 99%, pure
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
370080250 370081000
ACRO370080250EA 111 EUR
ACRO370080250 ACRO370081000
Zinc acetate 99%, pure
Zinc acetate
Formule: C₄H₆O₄Zn
Kookpunt: 908 °C (1013 hPa)
Smeltpunt: 83…86 °C
Dichtheid: 1,735 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012454
CAS nummer: 557-34-6
EINECS: 209-170-2
UN: 3077
ADR: 9,III
Merck Index: 12,10256
Order Now
Specification Test Results
Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Titration Complexometric ≥98.5 %