4-Aminobutyric acid

Leverancier: Chemicon
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CHMCAB5610EA 0 EUR
CHMCAB5610
4-Aminobutyric acid
4-Aminobutyric acid
Formule: C₄H₉NO₂
MW: 103,12 g/mol
Kookpunt: 248 °C (1013 hPa)
Smeltpunt: 202 °C
Dichtheid: 1,21 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008226
CAS nummer: 56-12-2
EINECS: 200-258-6
Merck Index: 13,00429

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR