Chemical resistant suit, Microchem® by AlphaTec® 5000, model 151-G02
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR50T-15108G02 OR50T-15102G02 OR50T-15103G02 OR50T-15104G02 OR50T-15105G02 OR50T-15106G02 OR50T-15107G02
ANSEOR50T-15108G02EA 1620 EUR
ANSEOR50T-15108G02 ANSEOR50T-15102G02 ANSEOR50T-15103G02 ANSEOR50T-15104G02 ANSEOR50T-15105G02 ANSEOR50T-15106G02 ANSEOR50T-15107G02
Chemical resistant suit, Microchem® by AlphaTec® 5000, model 151-G02
Wegwerpkleding
Highly visible rear entry suit, with neoprene rubber face seal for a close fit to full face respirators. Ideal for use in extremly hazardous areas where protection against concentrated chemicals and biological agents is required.
  • Neoprene rubber face seal
  • Rear horizontal zip entry
  • Attached socks with boot overflap
  • Attached Ansell Barrier gloves, with oversleeves and finger loops
  • Ultrasonically welded and taped seams provide a high barrier to liquid and particulates
  • Bright orange colour for improved worker safety
  • Multi-layer material is lightweight, yet strong and durable
Typical applications include: Industrial and tank cleaning, sewage purification installations, chemicals, oil and petrochemicals, pharmaceuticals, agriculture, HAZMAT Emergency Response (i.e. Level B), first response and fire service.
Certificaten: Type 3/4/5, EN 14126, EN 1073-2, EN 1149-5; Cat. III