Ethyl fluoroacetate ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Fluoroacetic ethyl ester
F0030-25ML F0030-500ML F0030-100ML
TCIAF0030-25MLEA 33 EUR
TCIAF0030-25ML TCIAF0030-500ML TCIAF0030-100ML
Ethyl fluoroacetate ≥98.0% (by GC)
Ethyl fluoroacetate
Formule: C₄H₇FO₂
MW: 106.1 g/mol
Kookpunt: 117°C
Vlampunt: 30°C
MDL Nummer: MFCD00000450
CAS nummer: 459-72-3
EINECS: 207-297-8
UN: 2929
ADR: 6.1 II
Merck Index: 14,04167
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a60f6b18b1a62b3e1358483c4370963ecd6bc70&vid=1a60f6b18b1a62b3e1358483c4370963ecd6bc70&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %