Difluoroacetic acid 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

162120025 162120100 162120500
ACRO162120025EA 42.4 EUR
ACRO162120025 ACRO162120100 ACRO162120500
Difluoroacetic acid 98%
Difluoroacetic acid
Formule: C₂H₂F₂O₂
Kookpunt: 132…134 °C (760 mmHg)
Smeltpunt: –1 °C
Dichtheid: 1,52 g/cm³
Vlampunt: 78 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004220
CAS nummer: 381-73-7
EINECS: 206-839-0
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.3420 to 1.3450 (20°C, 589 nm)
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR