Heptafluorobutyric acid for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Perfluorobutyric acid, HFBA, Heptafluorobutanoic acid
M-Clarity™ quality level = MQ200
8.43443.0025 8.43443.0005
8.43443.0025EA 87.8 EUR
8.43443.0025 8.43443.0005
Heptafluorobutyric acid for synthesis, Sigma-Aldrich®
Heptafluorobutyric acid
Formule: C₄HF₇O₂
MW: 214.04 g/mol
Kookpunt: 121
Smeltpunt: –17.5 °C
Dichtheid: 1.764 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004171
CAS nummer: 375-22-4
EINECS: 206-786-3
UN: 3265
ADR: 8 III