Trifluoroacetic anhydride ≥99%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Trifluoracetic anhydride, TFAA
A13614.0E
A13614.14
A13614.22
A13614.36
A13614.0EEA
767
EUR
A13614.0E
A13614.14
A13614.22
A13614.36
Trifluoroacetic anhydride ≥99%
Trifluoroacetic anhydride
Purity: 99+%
CAS No.: 407-25-0
Molecular Formula: C4F6O3
Formula Weight: 210.03
Storage Temperature: 2°C to 8°C
Refractive Index: 1.2690
Melting Point: -65°
Boiling Point: 39-40°
Beilstein Reference: 746197
UN No.: 3265
MDL No.: MFCD00000416
Formule:
C₄F₆O₃ MW: 210.03 g/mol Kookpunt: 39…40 °C Smeltpunt: –65 °C Dichtheid: 1.503 |
MDL Nummer:
MFCD00000416 CAS nummer: 407-25-0 EINECS: 206-982-9 UN: 3265 ADR: 8 I |